Learn More
Trajan Scientific Column HT5 QC3 Medical 1/EA
SDP
Catalog No. 5000494708
Encompass_Preferred
0 32 mm ID x 0 1 m film thickness x 12 m length HT5 GC capillary column
Catalog No. 50-004-94708 Supplier Trajan Scientific Supplier No. 054641
0 32 mm ID x 0 1 m film thickness x 12 m length HT5 GC capillary column