Learn More
Trajan Scientific Column HT5 QC3 Medical 1/EA
SDP
Catalog No. 5000493562
Encompass_Preferred
0 32 mm ID x 0 5 m film thickness x 15 m length HT5 GC capillary column
Catalog No. 50-004-93562 Supplier Trajan Scientific Supplier No. 054667
0 32 mm ID x 0 5 m film thickness x 15 m length HT5 GC capillary column