Learn More
Trajan Scientific Column HT5 QC5 Medical 1/EA
SDP
Supplier:  Trajan Scientific 054658
Encompass_Preferred
0 53 mm ID x 0 15 m film thickness x 25 m length HT5 GC capillary column
Catalog No. 50-004-93695