Learn More
Trajan Scientific Column HT5 QC5 Medical 1/EA
SDP
Catalog No. 5000494274
Encompass_Preferred
0 53 mm ID x 0 5 m film thickness x 30 m length HT5 GC capillary column
Catalog No. 50-004-94274 Supplier Trajan Scientific Supplier No. 054672
0 53 mm ID x 0 5 m film thickness x 30 m length HT5 GC capillary column