Learn More
Trajan Scientific Column HT8 QC2.5 Medical 1/EA
SDP
Catalog No. 5000493961
Encompass_Preferred
0 25 mm ID x 0 25 m film thickness x 30 m length HT8 GC capillary column
Catalog No. 50-004-93961 Supplier Trajan Scientific Supplier No. 054677
0 25 mm ID x 0 25 m film thickness x 30 m length HT8 GC capillary column