Learn More
Trajan Scientific Column HT8 QC2.5 PCB 1/EA
SDP
Catalog No. 5000494280
Encompass_Preferred
0 25 mm ID x 60 m length HT8-PCB GC capillary column
Catalog No. 50-004-94280 Supplier Trajan Scientific Supplier No. 054236
0 25 mm ID x 60 m length HT8-PCB GC capillary column