Learn More
Trajan Scientific Column HT8 QC3 Medical 1/EA
SDP
Catalog No. 5000493967
Encompass_Preferred
0 32 mm ID x 0 25 m film thickness x 25 m length HT8 GC capillary column
Catalog No. 50-004-93967 Supplier Trajan Scientific Supplier No. 054680
0 32 mm ID x 0 25 m film thickness x 25 m length HT8 GC capillary column