Learn More
Thermo Scientific COLUMN ID TAGS, 2PCS
Supplier:  Thermo Scientific 6732.0610
Encompass
Thermocouple temperature sensors for measuring and controlling temperature in the VH-C1 chromatography column by producing a voltage proportional to temperature difference.
Catalog No. NC1327438