Learn More
Thermo Scientific IonPac AS19 Column 2 x 250 mm
Supplier:  Thermo Scientific 083223
Encompass
2 x 250 mm analytical ion chromatography anion exchange column for inorganic anion separation using silica particles coated with latex and functionalized with quaternary ammonium
Catalog No. NC0899575