Learn More
Waters Corp Waters Corporation Tubing Assembly, U-Shaped, HPLC-Style Ferrule Seating, 3.9 in.
Catalog No. 50785904 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing assembly is a U-shaped column connector designed for efficient and error-minimized column joining in laboratory settings. It features end ferrules and nuts, with ferrules permanently seated to an HPLC-style depth setting, ensuring a precise connection for compatible hardware.
  • U-shaped column connector.
  • Includes end ferrules and nuts.
  • HPLC-style ferrule seating for compatible hardware.
  • Not for use with Parker-style or ACQUITY-based SEC columns.
  • Manufactured in ISO-certified plant using ultra-pure reagents.
  • Rigorous batch testing ensures consistent, reproducible performance.
  • Certified for optimal system performance and compliance.
  • Tubing length is approximately 3.9 in.
Catalog No. 50-785-904 Supplier Waters Corp Supplier No. WAT084080
This tubing assembly is a U-shaped column connector designed for efficient and error-minimized column joining in laboratory settings. It features end ferrules and nuts, with ferrules permanently seated to an HPLC-style depth setting, ensuring a precise connection for compatible hardware.
  • U-shaped column connector.
  • Includes end ferrules and nuts.
  • HPLC-style ferrule seating for compatible hardware.
  • Not for use with Parker-style or ACQUITY-based SEC columns.
  • Manufactured in ISO-certified plant using ultra-pure reagents.
  • Rigorous batch testing ensures consistent, reproducible performance.
  • Certified for optimal system performance and compliance.
  • Tubing length is approximately 3.9 in.