Learn More
Chiral Technologies, Inc. HPLC Column CHIRALCEL OD-3, 250 x 4.6 mm, 3 um
Catalog No. NC0259082
Encompass
Chiralcel Chiral Column, Particle Size: 3 um, Inner Diameter: 4.6mm, Length: 250mm.
Catalog No. NC0259082 Supplier Chiral Technologies, Inc. Supplier No. 14525
Chiralcel Chiral Column, Particle Size: 3 um, Inner Diameter: 4.6mm, Length: 250mm.