Learn More
Sigma Aldrich Fine Chemicals Biosciences Column PD-10 Empty Cytiva
Catalog No. 502904311
Encompass
Empty Disposable PD-10 Columns are economical plastic columns for single-use applications using gravity flow. The columns hold up to approx. 13 mL of chromatography media. The frits can be used as an upper bed support that prevents the medium from running dry under gravity flow.Together with LabMate Buffer Reservoir up to 25 mL of buffer and/or sample can be applied which reduces handling time considerably.
Catalog No. 50-290-4311 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GE17043501
Empty Disposable PD-10 Columns are economical plastic columns for single-use applications using gravity flow. The columns hold up to approx. 13 mL of chromatography media. The frits can be used as an upper bed support that prevents the medium from running dry under gravity flow.Together with LabMate Buffer Reservoir up to 25 mL of buffer and/or sample can be applied which reduces handling time considerably.