Learn More
LAMDA BIOTECH INC Column-Pure™ Animal Genomic DNA Kit
Catalog No. 502045800
Encompass_Preferred
This is a quick and easy spin column method for isolation of genomic DNA from various animal tissues and cultured cells. The kit is designed to process tissue samples up to 30mg. The isolated DNA is of high quality for many molecular biology applications, such as; PCR, restriction digestion, and other downstream applications.
Catalog No. 50-204-5800 Supplier LAMDA BIOTECH INC Supplier No. D427100
This is a quick and easy spin column method for isolation of genomic DNA from various animal tissues and cultured cells. The kit is designed to process tissue samples up to 30mg. The isolated DNA is of high quality for many molecular biology applications, such as; PCR, restriction digestion, and other downstream applications.