Learn More
LAMDA BIOTECH INC Column-Pure RNA Cleaning Conc
Supplier:  LAMDA BIOTECH INC D703
Encompass_Preferred
A spin column-based system designed for RNA purification and concentration from various sources. Suitable for cleaning RNA samples from in vitro transcription reactions and DNase treatments.
Catalog No. NC3726045