Learn More
LAMDA BIOTECH INC Column-Pure RNA Mini-Preps
Catalog No. NC3726044
Encompass_Preferred
A spin column-based system designed for total RNA isolation from animal tissues and cultured cells. The kit employs selective RNA binding to silica membrane technology for nucleic acid purification.
Catalog No. NC3726044 Supplier LAMDA BIOTECH INC Supplier No. D2312100
A spin column-based system designed for total RNA isolation from animal tissues and cultured cells. The kit employs selective RNA binding to silica membrane technology for nucleic acid purification.