Learn More
Chemglass Life Sciences Column, Chromatography, Quick Separation Funnel, 30mm Coarse Frit, 102mm height above the disc, 24/40 Lower Vacuum Assembly
SDP
Catalog No. CG141212
Encompass_Preferred
Column, Chromatography, Quick Separation Funnel, 30mm Coarse Frit, 102mm height above the disc, 24/40 Lower Vacuum Assembly. For performing quick separations using a vacuum source. All funnels have a lower standard taper inner joint with a sealed in drip tip and an outlet for connection to a vacuum source. All sizes have a Coarse porosity fritted disc. Special sizes are available, please call for details.
Catalog No. CG141212 Supplier Chemglass Life Sciences Supplier No. CG141212
Column, Chromatography, Quick Separation Funnel, 30mm Coarse Frit, 102mm height above the disc, 24/40 Lower Vacuum Assembly. For performing quick separations using a vacuum source. All funnels have a lower standard taper inner joint with a sealed in drip tip and an outlet for connection to a vacuum source. All sizes have a Coarse porosity fritted disc. Special sizes are available, please call for details.