Learn More
Teledyne ISCO, Inc RediSep Rf Silica Gel Disposable Flash Columns, 12 gram. Package of 20
Catalog No. 50871434 Shop All Teledyne ISCO Inc. Products
Encompass_Preferred
RediSep Rf Silica Gel Disposable Flash Columns, 12 gram. Package of 20. Disposable, bare silica column for Flash purification of organic compounds. The high quality media (40-60 µm particle size) is specified for high resolution and reproducibility for everyday purifications. Specifications:Reusability: Single use. Particle Size: 40-63 µm irregular. Mesh Size: 230-400.Pore Size: 60 Å. Surface Area m²/g: 500 ±50. Loading Capacity: 0.1-10%
Catalog No. 50-871-434 Supplier Teledyne ISCO, Inc Supplier No. 692203312
RediSep Rf Silica Gel Disposable Flash Columns, 12 gram. Package of 20. Disposable, bare silica column for Flash purification of organic compounds. The high quality media (40-60 µm particle size) is specified for high resolution and reproducibility for everyday purifications. Specifications:Reusability: Single use. Particle Size: 40-63 µm irregular. Mesh Size: 230-400.Pore Size: 60 Å. Surface Area m²/g: 500 ±50. Loading Capacity: 0.1-10%