Learn More
Teledyne ISCO, Inc RediSep Silver Silica Gel Disposable Flash Columns, 80 gram.  Package of 12.
Catalog No. 50871437 Shop All Teledyne ISCO Inc. Products
Encompass_Preferred
RediSep Rf disposable column for flash purification of organic compounds
Catalog No. 50-871-437 Supplier Teledyne ISCO, Inc Supplier No. 692203380
RediSep Rf disposable column for flash purification of organic compounds