Learn More
Pickering Laboratories Inc COLUMN SODIUM 4.6X110MM
SDP
Catalog No. NC0810879
Encompass_Preferred
30-minute High-efficiency Sodium Cation-exchange Column 4.6 x 110 mm, includes Amino Acid Test Mixture 1700-0070 Amino Acid Analysis.
Catalog No. NC0810879 Supplier Pickering Laboratories Inc Supplier No. 1154110T
Add to Cart
Add to Cart
30-minute High-efficiency Sodium Cation-exchange Column 4.6 x 110 mm, includes Amino Acid Test Mixture 1700-0070 Amino Acid Analysis.