Learn More
XENOSEP TECHNOLOGIES Sodium Sulfate 10G Columns, 48/pk
Catalog No. NC1906687
Encompass_Preferred
The XenoMax Sodium Sulfate Column is a prepacked column containing 10 grams of sodium sulfate which enables the efficient inline removal of water from the hexane extract as is passes from the SPE filter to the Flat Sided Flask.
Catalog No. NC1906687 Supplier XENOSEP TECHNOLOGIES Supplier No. 26524
The XenoMax Sodium Sulfate Column is a prepacked column containing 10 grams of sodium sulfate which enables the efficient inline removal of water from the hexane extract as is passes from the SPE filter to the Flat Sided Flask.