Learn More
Trajan Scientific Column Solgel QC2.5 1MS 1/EA
SDP
Catalog No. 5000494192
Encompass_Preferred
0 25 mm ID x 0 25 m film thickness x 60 m length SolGel-1ms GC capillary column
Catalog No. 50-004-94192 Supplier Trajan Scientific Supplier No. 054793
0 25 mm ID x 0 25 m film thickness x 60 m length SolGel-1ms GC capillary column