Learn More
Trajan Scientific Column Solgel QC2.5 1MS 1/EA
SDP
Supplier:  Trajan Scientific 054795
Encompass_Preferred
0 25 mm ID x 0 25 m film thickness x 30 m length SolGel-1ms GC capillary column
Catalog No. 50-004-93859