Learn More
Trajan Scientific Column Solgel QC2.5 Wax 1/EA
SDP
Catalog No. 5000494191
Encompass_Preferred
0 25 mm ID x 0 25 m film thickness x 30 m length SolGel-WAX GC capillary column
Catalog No. 50-004-94191 Supplier Trajan Scientific Supplier No. 054796
0 25 mm ID x 0 25 m film thickness x 30 m length SolGel-WAX GC capillary column