Learn More
Trajan Scientific Column Solgel QC2.5 Wax 1/EA
SDP
Supplier:  Trajan Scientific 054791
Encompass_Preferred
0 25 mm ID x 0 25 m film thickness x 60 m length SolGel-WAX GC capillary column
Catalog No. 50-004-93708