Learn More
Trajan Scientific Column Solgel QC2.5 Wax 1/EA
SDP
Catalog No. 5000493622
Encompass_Preferred
0 25 mm ID x 1 0 m film thickness x 30 m length SolGel-WAX GC capillary column
Catalog No. 50-004-93622 Supplier Trajan Scientific Supplier No. 054787
Add to Cart
Add to Cart
0 25 mm ID x 1 0 m film thickness x 30 m length SolGel-WAX GC capillary column