Learn More
Trajan Scientific Column Solgel QC3 1MS 1/EA
SDP
Catalog No. 5000494529
Encompass_Preferred
0 32 mm ID x 0 25 m film thickness x 30 m length SolGel-1ms GC capillary column
Catalog No. 50-004-94529 Supplier Trajan Scientific Supplier No. 054798
0 32 mm ID x 0 25 m film thickness x 30 m length SolGel-1ms GC capillary column