Learn More
Trajan Scientific Column Solgel QC3 Wax 1/EA
SDP
Catalog No. 5000494273
Encompass_Preferred
0 32 mm ID x 0 25 m film thickness x 30 m length SolGel-WAX GC capillary column
Catalog No. 50-004-94273 Supplier Trajan Scientific Supplier No. 054788
0 32 mm ID x 0 25 m film thickness x 30 m length SolGel-WAX GC capillary column