Learn More
Trajan Scientific Column Solgel QC3 Wax 1/EA
SDP
Catalog No. 5000494303
Encompass_Preferred
0 32 mm ID x 0 25 m film thickness x 60 m length SolGel-WAX GC capillary column
Catalog No. 50-004-94303 Supplier Trajan Scientific Supplier No. 054789
0 32 mm ID x 0 25 m film thickness x 60 m length SolGel-WAX GC capillary column