Learn More
Trajan Scientific Column Solgel QC3 Wax 1/EA
SDP
Catalog No. 5000493826
Encompass_Preferred
0 32 mm ID x 0 5 m film thickness x 30 m length SolGel-WAX GC capillary column on 5 cage
Catalog No. 50-004-93826 Supplier Trajan Scientific Supplier No. 054797CCS
0 32 mm ID x 0 5 m film thickness x 30 m length SolGel-WAX GC capillary column on 5 cage