Learn More
Trajan Scientific Column Solgel QC3 Wax 1/EA
SDP
Catalog No. 5000494331
Encompass_Preferred
0 32 mm ID x 0 5 m film thickness x 60 m length SolGel-WAX GC capillary column
Catalog No. 50-004-94331 Supplier Trajan Scientific Supplier No. 054792
0 32 mm ID x 0 5 m film thickness x 60 m length SolGel-WAX GC capillary column