Learn More
Trajan Scientific Column Solgel QC5 Wax 1/EA
SDP
Catalog No. 5000493614
Encompass_Preferred
0 53 mm ID x 0 5 m film thickness x 30 m length SolGel-WAX GC capillary column
Catalog No. 50-004-93614 Supplier Trajan Scientific Supplier No. 054786
0 53 mm ID x 0 5 m film thickness x 30 m length SolGel-WAX GC capillary column