Learn More
LAMDA BIOTECH INC Column-Pure™ Blood Genomic DNA Kit
Catalog No. 502045801
Encompass_Preferred
This is a quick and easy spin column method for isolation of genomic DNA from blood. The kit is designed to isolate genomic DNA from 300µl to 500μl whole blood. The purified genomic DNA is of suitable quality for many molecular biology applications, such as: PCR, restriction digestion, and other downstream applications.
Catalog No. 50-204-5801 Supplier LAMDA BIOTECH INC Supplier No. D483100
This is a quick and easy spin column method for isolation of genomic DNA from blood. The kit is designed to isolate genomic DNA from 300µl to 500μl whole blood. The purified genomic DNA is of suitable quality for many molecular biology applications, such as: PCR, restriction digestion, and other downstream applications.