Learn More
Mac-Mod ColumnSaver Pre-Column Filter, 0.5 um porosity, PEEK, (5/pk)
SDP
Catalog No. 5090002070
Encompass
Column Savers provide economical protection for HPLC columns packed with 3 m particle size or larger and with bore sizes (internal diameter) 3.0mm to 8.0mm. Use these Pre-column filters on the inlet of guard columns to extend the life and reduce the replacement costs of these items.
Catalog No. 50-900-02070 Supplier Mac-Mod Supplier No. MMCS205
Column Savers provide economical protection for HPLC columns packed with 3 m particle size or larger and with bore sizes (internal diameter) 3.0mm to 8.0mm. Use these Pre-column filters on the inlet of guard columns to extend the life and reduce the replacement costs of these items.