Learn More
Com-Pac International™ 3050 Infectious Substance Shipper
Description
Includes
This complete kit contains one 1-liter pressure vessel, bubble pouch and absorbent, two foam stabilizers, shipper's declarations, label and box. Increase the capacity of the Infecon 3050 by removing the cylindrical foam plug and adding another pressure vessel (Cat. No. 19-038-300C).
- Independently tested and certified
- Made in the U.S.A.
Specifications
Specifications
| Includes | This complete kit contains one 1-liter pressure vessel, bubble pouch and absorbent, two foam stabilizers, shipper's declarations, label and box. Increase the capacity of the Infecon 3050 by removing the cylindrical foam plug and adding another pressure vessel (Cat. No. 19-038-300C). |
| Certifications/Compliance | In compliance with all national and international regulations (DOT and ICAO), the Infecon 3050 also complies with the Federal Express™ IATA Operator Variation identified in section 2.9.3. The FX-11 regulation requires all packaging to accommodate the Federal Express and regulatory documentation as well as all regulatory marketing and labeling on the top and sides of the box. |
| Type | RT Infectious Substance Shipper |