Learn More
Cayman Chemical Coomassie Blue R-250, CAS: 6104-59-2, 25 g
SDP
Catalog No. 502971652
Encompass_Preferred
A dye that is commonly used in laboratories to stain or quantify proteins; typically gives blue bands on a clear background with a sensitivity of 50-100 ng/band; displays metachromasia by appearing pink-red, rather than blue, when binding certain proteins, such as collagen and histone H1
Catalog No. 50-297-1652 Supplier Cayman Chemical Supplier No. 1450025 G
A dye that is commonly used in laboratories to stain or quantify proteins; typically gives blue bands on a clear background with a sensitivity of 50-100 ng/band; displays metachromasia by appearing pink-red, rather than blue, when binding certain proteins, such as collagen and histone H1