Learn More
Cayman Chemical ComasIe Blue R-250 50g
SDP
Catalog No. 502972844
Encompass_Preferred
A dye that is commonly used in laboratories to stain or quantify proteins; typically gives blue bands on a clear background with a sensitivity of 50-100 ng/band; displays metachromasia by appearing pink-red, rather than blue, when binding certain proteins, such as collagen and histone H1
Catalog No. 50-297-2844 Supplier Cayman Chemical Supplier No. 1450050 G
A dye that is commonly used in laboratories to stain or quantify proteins; typically gives blue bands on a clear background with a sensitivity of 50-100 ng/band; displays metachromasia by appearing pink-red, rather than blue, when binding certain proteins, such as collagen and histone H1