Learn More
Bio Rad Laboratories COMB 10-WELL
Catalog No. 509011138
Encompass
Comb, 10-Well; Bio-Rad; 14cm wide, 0.75 thick, adjustable ht.; Combs for sub-cell, Wide mini-sub cell electrophoresis cells
Catalog No. 50-901-1138 Supplier Bio Rad Laboratories Supplier No. 1704325
Comb, 10-Well; Bio-Rad; 14cm wide, 0.75 thick, adjustable ht.; Combs for sub-cell, Wide mini-sub cell electrophoresis cells