Learn More
Bio Rad Laboratories COMB 30-WELL
Catalog No. 509010263
Encompass
Comb; Bio-Rad; For Sub-Cell and Wide Mini-Sub cell electrophoresis cells; 30-Well; 14cm wide; 1.5mm thick; Adjustable height
Catalog No. 50-901-0263 Supplier Bio Rad Laboratories Supplier No. 1704344
Comb; Bio-Rad; For Sub-Cell and Wide Mini-Sub cell electrophoresis cells; 30-Well; 14cm wide; 1.5mm thick; Adjustable height