Learn More
Sensorex COMB PH ELCTRDE FLAT-BNC CNCTR
Catalog No. NC9329593
Encompass_Preferred
Combination pH/Reference Electrodes with EPOXY BODIES,Standard Laboratory use,GLASS TYPE:Flat,JUNCTION:HDPE/fiber,SIZE:DIA :9/16 in. (15mm);LENGTH :4 1/2 in. (115mm),SPECIAL FEATURES:Flat Surface,Double Junction,sealed,gel-filled reference
Catalog No. NC9329593 Supplier Sensorex Supplier No. S450CD
Combination pH/Reference Electrodes with EPOXY BODIES,Standard Laboratory use,GLASS TYPE:Flat,JUNCTION:HDPE/fiber,SIZE:DIA :9/16 in. (15mm);LENGTH :4 1/2 in. (115mm),SPECIAL FEATURES:Flat Surface,Double Junction,sealed,gel-filled reference