Learn More
Hach Company Radiometer Analytical PHC3001-9 Combination pH Electrode (glass body, Ag/AgCl ref., screw cap)
Catalog No. 501997573 Shop All Hach Company Products
Encompass
The Radiometer PHC3001-9 Combination pH Electrode has a glass body, Ag/AgCl reference system, refillable electrolyte (3M KCl + sat. AgCl) and porous pin junction for general purpose applications. The PHC3001-9 is a screw cap electrode and requires the CL114 cable (part no. A94L114) for compatibilty with Hach Sension+ laboratory pH meters (or any meter with a BNC connection).
Catalog No. 50-199-7573 Supplier Hach Company Supplier No. E16M301
The Radiometer PHC3001-9 Combination pH Electrode has a glass body, Ag/AgCl reference system, refillable electrolyte (3M KCl + sat. AgCl) and porous pin junction for general purpose applications. The PHC3001-9 is a screw cap electrode and requires the CL114 cable (part no. A94L114) for compatibilty with Hach Sension+ laboratory pH meters (or any meter with a BNC connection).