Learn More
Hach Company Radiometer Analytical PHC3005-9 Combination pH Electrode (epoxy body, Ag/AgCl ref., screw cap)
Catalog No. 501997575 Shop All Hach Company Products
Encompass
The Radiometer PHC3005-9 Combination pH Electrode has a robust epoxy body, Ag/AgCl reference system, refillable electrolyte (3M KCl + sat. AgCl) and porous pin junction for general purpose applications. The PHC3005-9 is a screw cap electrode and requires the CL114 cable (part no. A94L114) for compatibilty with Hach Sension+ laboratory pH meters (or any meter with a BNC connection).
Catalog No. 50-199-7575 Supplier Hach Company Supplier No. E16M303
The Radiometer PHC3005-9 Combination pH Electrode has a robust epoxy body, Ag/AgCl reference system, refillable electrolyte (3M KCl + sat. AgCl) and porous pin junction for general purpose applications. The PHC3005-9 is a screw cap electrode and requires the CL114 cable (part no. A94L114) for compatibilty with Hach Sension+ laboratory pH meters (or any meter with a BNC connection).