Learn More
Hach Company Comb pH incer. sens PVDF flat
Catalog No. 5000207790 Shop All Hach Company Products
Encompass
Digital 3/4in Combination pH/ORP Sensor-Insertion Style-PVDF Body Material-Flat Glass pH ElectrodePVDF Insertion Body Style 4 5m Analog Cable Flat Glass pH Electrode Pt 1000 Temperature Compensation includes 6120600 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3ft) Digital Extension Cable
Catalog No. 50-002-07790 Supplier Hach Company Supplier No. DPC2K2A
Add to Cart
Add to Cart
Digital 3/4in Combination pH/ORP Sensor-Insertion Style-PVDF Body Material-Flat Glass pH ElectrodePVDF Insertion Body Style 4 5m Analog Cable Flat Glass pH Electrode Pt 1000 Temperature Compensation includes 6120600 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3ft) Digital Extension Cable