Learn More
Hach Company Comb pH San. sens SS/PVDF gls
Catalog No. 5000208048 Shop All Hach Company Products
Encompass
Digital 3/4in Combination pH/ORP Sensor-Sanitary Sensor Style-316 SS/PVDF Body Material-General Purpose Glass pH Electrode316 SSVDF Sanitary Body Style 4 5m Analog Cable General Purpose Glass pH Electrode Pt 1000 Temperature Compensation includes 6120600 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3ft) Digital Extension Cable
Catalog No. 50-002-08048 Supplier Hach Company Supplier No. DPC3K2A
Digital 3/4in Combination pH/ORP Sensor-Sanitary Sensor Style-316 SS/PVDF Body Material-General Purpose Glass pH Electrode316 SSVDF Sanitary Body Style 4 5m Analog Cable General Purpose Glass pH Electrode Pt 1000 Temperature Compensation includes 6120600 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3ft) Digital Extension Cable