Learn More
Hach Company Comb pH sensor PPS flat glass
Catalog No. 5000207716 Shop All Hach Company Products
Encompass
GLI Digital 3/4 Inch Combination pH/ORP Sensor-Convertible Style-PPS Body-Flat Glass ElectrodePPS Convertible Body Style 4 5m Analog Cable General Purpose Glass pH Electrode No Temperature Compensation includes 6120600 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3 ft) Digital Extension Cable
Catalog No. 50-002-07716 Supplier Hach Company Supplier No. DPC1R1N
GLI Digital 3/4 Inch Combination pH/ORP Sensor-Convertible Style-PPS Body-Flat Glass ElectrodePPS Convertible Body Style 4 5m Analog Cable General Purpose Glass pH Electrode No Temperature Compensation includes 6120600 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3 ft) Digital Extension Cable