Learn More
Hach Company Comb pH sensor PPS glass
Catalog No. 5000207678 Shop All Hach Company Products
Encompass
Digital 3/4in Combination pH Sensor-Convertible Sensor Style-PPS Body Material-General Purpose Glass pH ElectrodePPS Convertible Body Style 4 5m Analog Cable General Purpose Glass pH Electrode Pt 1000 Temperature Compensation Cold Temperature Operation Fill includes 6120600 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3ft) Digital Extension Cable
Catalog No. 50-002-07678 Supplier Hach Company Supplier No. DPC1R1A
Add to Cart
Add to Cart
Digital 3/4in Combination pH Sensor-Convertible Sensor Style-PPS Body Material-General Purpose Glass pH ElectrodePPS Convertible Body Style 4 5m Analog Cable General Purpose Glass pH Electrode Pt 1000 Temperature Compensation Cold Temperature Operation Fill includes 6120600 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3ft) Digital Extension Cable