Learn More
Hach Company Comb pH sensor PPS HF-Res gls
Catalog No. 5000207717 Shop All Hach Company Products
Encompass
Digital 3/4in Combination pH/ORP Sensor-Convertible Style-PPS Body-HF-Resistant Glass pH ElectrodePPS Convertible Body Style 4 5m Analog Cable HF-resistant Glass pH Electrode Pt 1000 Temperature Compensation includes 6120600 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3 ft) Digital Extension Cable
Catalog No. 50-002-07717 Supplier Hach Company Supplier No. DPC1R3A
Digital 3/4in Combination pH/ORP Sensor-Convertible Style-PPS Body-HF-Resistant Glass pH ElectrodePPS Convertible Body Style 4 5m Analog Cable HF-resistant Glass pH Electrode Pt 1000 Temperature Compensation includes 6120600 Digital Gateway for Connecting to sc Controller and a 6122400 1m (3 ft) Digital Extension Cable