Learn More
MSC Rochester Midland Heavy Duty Degreaser, Citrus, Concentrated, 5 gal
Catalog No. 502817210
Encompass
Combat is a citrus-based heavy duty degreaser suitable for a wide variety of applications. It features a safe, unique formula that effectively cuts through heavy grease, oil, and other factory solids, and also dissolves rubber tire marks on concrete.
  • Imparts a fresh, citrus odor to the workplace.
  • Quickly penetrates oils, greases, shop soils, and impacted grime.
  • Free rinsing, leaving a clean, soil-free surface.
  • Safe on aluminum, stainless steel, rubber, and plastic.
  • Superior oil/solution separation.
  • Low foaming, suitable for recirculating wash equipment and automatic floor scrubbers.
Catalog No. 50-281-7210 Supplier MSC Supplier No. 90709395
Combat is a citrus-based heavy duty degreaser suitable for a wide variety of applications. It features a safe, unique formula that effectively cuts through heavy grease, oil, and other factory solids, and also dissolves rubber tire marks on concrete.
  • Imparts a fresh, citrus odor to the workplace.
  • Quickly penetrates oils, greases, shop soils, and impacted grime.
  • Free rinsing, leaving a clean, soil-free surface.
  • Safe on aluminum, stainless steel, rubber, and plastic.
  • Superior oil/solution separation.
  • Low foaming, suitable for recirculating wash equipment and automatic floor scrubbers.