Learn More
eMolecules​ 22139-77-1 | Pinosylvin | Combi-Blocks, Inc. | MFCD00210544 | 212.248 | C14H12O2 | 98.000 | Oc1cc(O)cc(\C=C\c2ccccc2)c1 | 250mg | 603236210
Catalog No. 5000127452 Shop All eMolecules Inc. Products
Encompass_Preferred
Pinosylvin | Combi-Blocks, Inc. | 22139-77-1 | MFCD00210544 | 212.248 | C14H12O2 | 98.000 | Oc1cc(O)cc(\C=C\c2ccccc2)c1 | 250mg | 603236210
Catalog No. 50-001-27452 Supplier eMolecules​ Supplier No. 603236210
Pinosylvin | Combi-Blocks, Inc. | 22139-77-1 | MFCD00210544 | 212.248 | C14H12O2 | 98.000 | Oc1cc(O)cc(\C=C\c2ccccc2)c1 | 250mg | 603236210