Learn More
eMolecules​ 98-92-0 | Nicotinamide | Combi-Blocks | MFCD00006395 | 122.127 | C6H6N2O | 98.000 | NC(=O)c1cccnc1 | 1kg | 457916668
Catalog No. 5000129660 Shop All eMolecules Inc. Products
Encompass_Preferred
Nicotinamide | Combi-Blocks | 98-92-0 | MFCD00006395 | 122.127 | C6H6N2O | 98.000 | NC(=O)c1cccnc1 | 1kg | 457916668
Catalog No. 50-001-29660 Supplier eMolecules​ Supplier No. 457916668
Nicotinamide | Combi-Blocks | 98-92-0 | MFCD00006395 | 122.127 | C6H6N2O | 98.000 | NC(=O)c1cccnc1 | 1kg | 457916668