Learn More
HANNA INSTRUMENTS INC Refillable Combination pH Electrode with BNC Connector
SDP
Catalog No. NC1882407
Encompass
Perfect for general laboratory use, the HI1131B is a glass-bodied combination pH electrode with a BNC connector for a quick and easy connection to any BNC pH meter. With a double junction design and the ability to change out contaminated electrolyte, this economical pH electrode ensures great results.Easy to clean glass body for faster measurements.BNC connector for quick and easy connection to most pH meters.Great for general laboratory use.
Catalog No. NC1882407 Supplier HANNA INSTRUMENTS INC Supplier No. HI1131B
Perfect for general laboratory use, the HI1131B is a glass-bodied combination pH electrode with a BNC connector for a quick and easy connection to any BNC pH meter. With a double junction design and the ability to change out contaminated electrolyte, this economical pH electrode ensures great results.Easy to clean glass body for faster measurements.BNC connector for quick and easy connection to most pH meters.Great for general laboratory use.